2-(3-Methylphenoxy)ethanol, 98%, Thermo Scientific Chemicals
2-(3-Methylphenoxy)ethanol, 98%, Thermo Scientific Chemicals
2-(3-Methylphenoxy)ethanol, 98%, Thermo Scientific Chemicals
2-(3-Methylphenoxy)ethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(3-Methylphenoxy)ethanol, 98%, Thermo Scientific Chemicals

Quantity:
2 g
10 g
Catalog number L04984.09
also known as L04984-09
Price (JPY)
-
Request A Quote
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS13605-19-1
IUPAC Name2-(3-methylphenoxy)ethan-1-ol
Molecular FormulaC9H12O2
InChI KeyFDBXUXVQIOQYIX-UHFFFAOYSA-N
SMILESCC1=CC=CC(OCCO)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
Assay (GC)>97.5%
FormLiquid
Refractive Index1.5300-1.5340 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only