Ethyl butyrate, 99%
Ethyl butyrate, 99%
Ethyl butyrate, 99%
Thermo Scientific Chemicals

Ethyl butyrate, 99%

CAS: 105-54-4 | C6H12O2 | 116.16 g/mol
Quantity:
250 mL
1000 mL
Catalog number L06025.AK
also known as L06025-AK
Price (JPY)
-
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS105-54-4
IUPAC Nameethyl butanoate
Molecular FormulaC6H12O2
InChI KeyOBNCKNCVKJNDBV-UHFFFAOYSA-N
SMILESCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.3910-1.3940 @ 20?C
FormLiquid
CommentClear colorless
Ethyl butyrate is used in flavors and fragrances. It is used as a solvent in perfumery products, and as a plasticizer for cellulose. It is used in the preparation of novel 2-cyanopyrimidines as cathespin K inhibitors. It is also used in the synthesis of pyridobenzimidazole derivatives exhibiting antifungal activity by the inhibition of β-1,6-glucan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl butyrate is used in flavors and fragrances. It is used as a solvent in perfumery products, and as a plasticizer for cellulose. It is used in the preparation of novel 2-cyanopyrimidines as cathespin K inhibitors. It is also used in the synthesis of pyridobenzimidazole derivatives exhibiting antifungal activity by the inhibition of β-1,6-glucan.

Solubility
Soluble in propylene glycol, paraffin oil, and kerosene.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only

General References:

  1. Kaitlin S Carlson; Maggie R Dillione; Daniel W Wesson. Odor- and state-dependent olfactory tubercle local field potential dynamics in awake rats. Journal of Neurophysiology.2014, 111, (10), 2109-2123.
  2. P Sampranpiboon; R Jiraratananon; D Uttapap; X Feng; R.Y.M Huang. Pervaporation separation of ethyl butyrate and isopropanol with polyether block amide (PEBA) membranes. Journal of Membrane Science.2000, 173, (10), 53-59.