3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98+%
3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98+%
3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98+%
Thermo Scientific Chemicals

3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98+%

CAS: 2455-14-3 | C28H40O2 | 408.626 g/mol
Quantity:
2 g
10 g
Catalog number L07743.09
also known as L07743-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS2455-14-3
IUPAC Name3,3',5,5'-tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione
Molecular FormulaC28H40O2
InChI KeyGQIGHOCYKUBBOE-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(C=C(C1=O)C(C)(C)C)=C1C=C(C(=O)C(=C1)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Formcrystals
Appearance (Color)Dark red-purple
Assay (HPLC)> 98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only