2-Chlorobenzoxazole, 98%
2-Chlorobenzoxazole, 98%
2-Chlorobenzoxazole, 98%
2-Chlorobenzoxazole, 98%
Thermo Scientific Chemicals

2-Chlorobenzoxazole, 98%

CAS: 615-18-9 | C7H4ClNO | 153.57 g/mol
Quantity:
5 g
25 g
Catalog number L07981.14
also known as L07981-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS615-18-9
IUPAC Name2-chloro-1,3-benzoxazole
Molecular FormulaC7H4ClNO
InChI KeyBBVQDWDBTWSGHQ-UHFFFAOYSA-N
SMILESClC1=NC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.5650-1.5690 @ 20?C
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥97.5%
2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible in water.

Notes
Store under inert gas. Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Keep away from heat.
RUO – Research Use Only

General References:

  1. J Sam; J L Valentine; C W Richmond. Reaction of 2-chlorobenzoxazole with anthranilic acids and 2-amino-1-naphthalenesulfonic acid.Journal of Pharmaceutical Sciences.1968, 57 (8), 1445-1446.
  2. Francesco Babudri; Saverio Florio; Ludovico Ronzini. Cross-coupling of 2-chlorobenzoxazole with β,γ -unsaturated grignard reagents: regioselective synthesis of 2-allyl-, 2-allenyl-, and 2-propargyl-benzoxazoles.Tetrahedron.1986, 42 3905-3911.