tert-Butyl 2-bromoisobutyrate, 97%
tert-Butyl 2-bromoisobutyrate, 97%
tert-Butyl 2-bromoisobutyrate, 97%
tert-Butyl 2-bromoisobutyrate, 97%
Thermo Scientific Chemicals

tert-Butyl 2-bromoisobutyrate, 97%

CAS: 23877-12-5 | C8H15BrO2 | 223.11 g/mol
Quantity:
10 g
50 g
250 g
Catalog number L08216.09
also known as L08216-09
Price (JPY)
-
Request A Quote
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS23877-12-5
IUPAC Nametert-butyl 2-bromo-2-methylpropanoate
Molecular FormulaC8H15BrO2
InChI KeyIGVNJALYNQVQIT-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)C(C)(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4340-1.4390 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only