2,2,2-Trimethylacetophenone, 97%
2,2,2-Trimethylacetophenone, 97%
2,2,2-Trimethylacetophenone, 97%
Thermo Scientific Chemicals

2,2,2-Trimethylacetophenone, 97%

CAS: 938-16-9 | C11H14O | 162.232 g/mol
Quantity:
1 g
5 g
25 g
Catalog number L08729.14
also known as L08729-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS938-16-9
IUPAC Name2,2-dimethyl-1-phenylpropan-1-one
Molecular FormulaC11H14O
InChI KeyOECPUBRNDKXFDX-UHFFFAOYSA-N
SMILESCC(C)(C)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5065-1.5105 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Not miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
RUO – Research Use Only
O. Scialdone; M.A. Sabatino; C. Belfiore; A. Galia; M.P. Paternostro; G. Filardo; An unexpected ring carboxylation in the electrocarboxylation of aromatic ketones.Electrochimica Acta.2006, 51 3500-3505.