3,3'-Diaminobenzophenone, 90%
3,3'-Diaminobenzophenone, 90%
3,3'-Diaminobenzophenone, 90%
Thermo Scientific Chemicals

3,3'-Diaminobenzophenone, 90%

CAS: 611-79-0 | C13H12N2O | 212.25 g/mol
Quantity:
1 g
5 g
Catalog number L09128.06
also known as L09128-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS611-79-0
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥88.0%
Appearance (Color)Yellow to pale brown
Assay (Non-aqueous acid-base Titration)≥88.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only