N-(4-Nitrophenyl)thiourea, 98%
N-(4-Nitrophenyl)thiourea, 98%
N-(4-Nitrophenyl)thiourea, 98%
Thermo Scientific Chemicals

N-(4-Nitrophenyl)thiourea, 98%

CAS: 3696-22-8 | C7H7N3O2S | 197.21 g/mol
Quantity:
1 g
5 g
Catalog number L09331.06
also known as L09331-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3696-22-8
IUPAC Name(4-nitrophenyl)thiourea
Molecular FormulaC7H7N3O2S
InChI KeyBLYAANPIHFKKMQ-UHFFFAOYSA-N
SMILESNC(=S)NC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown to gray
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only