Tetramethoxysilane, 98%
Tetramethoxysilane, 98%
Tetramethoxysilane, 98%
Thermo Scientific Chemicals

Tetramethoxysilane, 98%

CAS: 681-84-5 | C4H12O4Si | 152.22 g/mol
Quantity:
25 g
100 g
500 g
Catalog number L09716.22
also known as L09716-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS681-84-5
IUPAC Nametetramethyl silicate
Molecular FormulaC4H12O4Si
InChI KeyLFQCEHFDDXELDD-UHFFFAOYSA-N
SMILESCO[Si](OC)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5% (UK Sourced Material)
Identification (FTIR)Conforms (UK Sourced Material)
Appearance (Color)Clear colorless
FormLiquid
CommentMaterial Sourced in UK and US
View more
Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

Solubility
Miscible with ethanol and organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, acids and bases.
RUO – Research Use Only

General References:

  1. Han, C.; Li, M. Y.; Li, Y. N.; Liu, H. L.; Wang, P.; Collinson, M. M.; Lu, Z. X. Self-supporting hybrid silica membranes with 3D large-scale ordered interconnected pore architectures. RSC Adv. 2015, 5 (25), 19182-19189.
  2. Yao, D.; Chen, Y.; Jin, R. Different dimensional silica materials prepared using shaped block copolymer nanoobjects as catalytic templates. J. Mater. Chem. B 2015, 3 (28), 5786-5794.