Dimethyl adipimidate dihydrochloride was used as a cross-linking reagent in the study of rhodopsin in sheep photoreceptor membrane
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl adipimidate dihydrochloride was used as a cross-linking reagent in the study of rhodopsin in sheep photoreceptor membrane
Solubility
Hydrolyses in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, water.
RUO – Research Use Only
T. H. Ji. Crosslinking of the Glycoproteins in Human Erythrocyte Membranes PNAS. 1974, 71 (1), 93-95.
Investigation of the organization of rhodopsin in the sheep photoreceptor membrane by using cross-linking reagents. BioChem.: Indian J. 1979, 177 215-223.
Cross-linking agent for modification of proteins via amidation.