Thermo Scientific Chemicals

Hexafluoroacetone trihydrate, 98%, Thermo Scientific Chemicals

Catalog number: L10777.06
5 g, Each
Thermo Scientific Chemicals

Hexafluoroacetone trihydrate, 98%, Thermo Scientific Chemicals

Catalog number: L10777.06
5 g, Each
Quantity
Catalog number: L10777.06
also known as L10777-06
Price (USD)
Price: 36.70
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
34202-69-2
IUPAC Name
hexafluoropropan-2-one trihydrate
Molecular Formula
C3H6F6O4
InChI Key
SNZAEUWCEHDROX-UHFFFAOYSA-N
SMILES
O.O.O.FC(F)(F)C(=O)C(F)(F)F
Refractive Index
1.3160-1.3220 @ 20°C
Form
Liquid
Water Content (Karl Fischer Titration)
21.0-30.0% (2.4-3.4 waters of hydration)
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text