1-Dimethylamino-2-propyne, 98%
1-Dimethylamino-2-propyne, 98%
1-Dimethylamino-2-propyne, 98%
Thermo Scientific Chemicals

1-Dimethylamino-2-propyne, 98%

CAS: 7223-38-3 | C5H9N | 83.134 g/mol
Quantity:
5 g
25 g
Catalog number L12336.14
also known as L12336-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7223-38-3
IUPAC Namedimethyl(prop-2-yn-1-yl)amine
Molecular FormulaC5H9N
InChI KeyILBIXZPOMJFOJP-UHFFFAOYSA-N
SMILESCN(C)CC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.4175-1.4215 @ 20?C
FormLiquid
1-Dimethylamino-2-propyne is used in the synthesis of core shell systems based on noble metal nanoparticles coated by copolymers (polyacetylenes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dimethylamino-2-propyne is used in the synthesis of core shell systems based on noble metal nanoparticles coated by copolymers (polyacetylenes).

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to carbon dioxide. Store under inert gas.
RUO – Research Use Only

General References:

  1. Ilaria Fratoddi, Iole Venditti.; Chiara Battocchio.; Giovanni Polzonetti.; Cesare Cametti.; Maria Vittoria Russo. Core shell hybrids based on noble metal nanoparticles and conjugated polymers: synthesis and characterization. Nanoscale Research Letters. 2011, 6 (1), 98.
  2. Ledlie DB.; Miller G. 3-(Dimethylamino)-1-propyne: convenient precursor for a versatile mixed cuprate reagent. J. Org. Chem. 1979, 44 (6), 1006-1007.