2-Bromophenyl isocyanate, 97%
2-Bromophenyl isocyanate, 97%
2-Bromophenyl isocyanate, 97%
2-Bromophenyl isocyanate, 97%
Thermo Scientific Chemicals

2-Bromophenyl isocyanate, 97%

CAS: 1592-00-3 | C7H4BrNO | 198.019 g/mol
Quantity:
1 g
5 g
25 g
Catalog number L12521.14
also known as L12521-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1592-00-3
IUPAC Name1-bromo-2-isocyanatobenzene
Molecular FormulaC7H4BrNO
InChI KeyGOOVAYJIVMBWPP-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.5820-1.5870 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only