5,5-Dimethylhexane-2,4-dione, 99%
5,5-Dimethylhexane-2,4-dione, 99%
Thermo Scientific Chemicals

5,5-Dimethylhexane-2,4-dione, 99%

CAS: 7307-04-2 | C9H9NO4 | 195.17 g/mol
Quantity:
1 g
5 g
Catalog number L13104.06
also known as L13104-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7307-04-2
IUPAC Name5,5-dimethylhexane-2,4-dione
Molecular FormulaC8H14O2
InChI KeyLCLCVVVHIPPHCG-UHFFFAOYSA-N
SMILESCC(=O)CC(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.4570-1.4610 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only