5(6)-Carboxyfluorescein, mixture of isomers, 97%
5(6)-Carboxyfluorescein, mixture of isomers, 97%
5(6)-Carboxyfluorescein, mixture of isomers, 97%
Thermo Scientific Chemicals

5(6)-Carboxyfluorescein, mixture of isomers, 97%

CAS: 72088-94-9 | C42H24O14 | 752.64 g/mol
Quantity:
250 mg
1 g
Catalog number L13439.03
also known as L13439-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS72088-94-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown or orange-red or orange-brown
FormPowder or crystalline powder and/or chunks
Assay (HPLC)≥96.0% (sum of isomers)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only