Thermo Scientific Chemicals

Dowex™ 1X8 100-200 (Cl), Thermo Scientific Chemicals

Catalog number: L14257.18
50 g, Each
Thermo Scientific Chemicals

Dowex™ 1X8 100-200 (Cl), Thermo Scientific Chemicals

Catalog number: L14257.18
50 g, Each
Quantity
Catalog number: L14257.18
also known as L14257-18
Price (USD)
Price: 46.70
Online price: 39.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12627-85-9
SMILES
*
MDL Number
MFCD00132718
Appearance (Color)
White to pale yellow or pale cream
Matrix
Trimethylbenzylammonium active functional group
Exchange capacity
≥1.2 eq/L (Cl)
Moisture holding capacity
39-45% (Cl)
Particle Size
>250 micron : ≤15.0%

Description

Dowex™ 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text