delta-Dodecanolactone, 98%
delta-Dodecanolactone, 98%
delta-Dodecanolactone, 98%
delta-Dodecanolactone, 98%
Thermo Scientific Chemicals

delta-Dodecanolactone, 98%

CAS: 713-95-1 | C12H22O2 | 198.306 g/mol
Quantity:
25 g
100 g
Catalog number L14406.14
also known as L14406-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS713-95-1
IUPAC Name6-heptyloxan-2-one
Molecular FormulaC12H22O2
InChI KeyQRPLZGZHJABGRS-UHFFFAOYNA-N
SMILESCCCCCCCC1CCCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4580-1.4620 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
delta-Dodecanolactone is used as a :flavoring agent. It is used in daily chemical essence. It is also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Dodecanolactone is used as a :flavoring agent. It is used in daily chemical essence. It is also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas.
RUO – Research Use Only