Thermo Scientific Chemicals

N,N-Dimethylcyclohexylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L14521.AE
100 mL, Each
Thermo Scientific Chemicals

N,N-Dimethylcyclohexylamine, 98+%, Thermo Scientific Chemicals

Catalog number: L14521.AE
100 mL, Each
Quantity
Catalog number: L14521.AE
also known as L14521-AE
Price (USD)
Price: 49.70
Online price: 42.65
Your price:
Quantity
-

Chemical Identifiers

CAS
98-94-2
IUPAC Name
N,N-dimethylcyclohexanamine
Molecular Formula
C8H17N
InChI Key
SVYKKECYCPFKGB-UHFFFAOYSA-N
SMILES
CN(C)C1CCCCC1
Refractive Index
1.4520-1.4560 @ 20?C
Assay (GC)
≥98.0%
Form
Liquid
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text