Thermo Scientific Chemicals

m-Tolyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: L14602.22
100 g, Each
Thermo Scientific Chemicals

m-Tolyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: L14602.22
100 g, Each
Quantity
Catalog number: L14602.22
also known as L14602-22
Price (USD)
Quantity
-

Description

m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text