Benzimidazole-5,6-dicarboxylic acid, 97%
Benzimidazole-5,6-dicarboxylic acid, 97%
Benzimidazole-5,6-dicarboxylic acid, 97%
Thermo Scientific Chemicals

Benzimidazole-5,6-dicarboxylic acid, 97%

CAS: 10351-75-4 | C9H6N2O4 | 206.16 g/mol
Quantity:
1 g
5 g
Catalog number L14754.03
also known as L14754-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS10351-75-4
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Benzimidazole-5,6-dicarboxylic acid is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole-5,6-dicarboxylic acid is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Xue Ma.; Xia Li.; Yu-E Cha.; Lin-Pei Jin. Highly Thermostable One-Dimensional Lanthanide(III) Coordination Polymers Constructed from Benzimidazole-5,6-dicarboxylic Acid and 1,10-Phenanthroline: Synthesis, Structure, and Tunable White-Light Emission. Cryst. Growth Des. 201'2, 12 (11), 5227-5232.
  2. Zhao-Xi Wang.; Qiong-Fang Wu.; Hong-Jiang Liu.; Min Shao.; Hong-Ping Xiao.; Ming-Xing Li. 2D and 3D lanthanide coordination polymers constructed from benzimidazole-5,6-dicarboxylic acid and sulfate bridged secondary building units. CrystEngComm. 2010, 12 1139-1146.