1-Ethylpiperazine, 98%
1-Ethylpiperazine, 98%
1-Ethylpiperazine, 98%
Thermo Scientific Chemicals

1-Ethylpiperazine, 98%

CAS: 5308-25-8 | C6H14N2 | 114.192 g/mol
Quantity:
25 g
100 g
500 g
Catalog number L15539.14
also known as L15539-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5308-25-8
IUPAC Name1-ethylpiperazine
Molecular FormulaC6H14N2
InChI KeyWGCYRFWNGRMRJA-UHFFFAOYSA-N
SMILESCCN1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.4670-1.4710 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
1-Ethylpiperazine is used in the synthesis of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride. It is also employed as an intermediate in veterinary medicine, especially in the production of ciproflmoxacin ethyl. Further, it serves as a precursor to prepare dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethylpiperazine is used in the synthesis of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride. It is also employed as an intermediate in veterinary medicine, especially in the production of ciproflmoxacin ethyl. Further, it serves as a precursor to prepare dyes.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kalia, S. B.; Kumar, D.; Sharma, M.; Christopher, J. Physico-chemical investigations and thermal degradation behaviour of 4-ethylpiperazine-1-carbodithioic acid complexes of cobalt(II). J. Therm. Anal. Calorim. 2015, 120 (2), 1099-1106.
  2. Kalia, S. B.; Puri, R.; Thakur, A.; Christopher, J. Synthesis, characterization and thermal degradation studies on some oxovanadium(IV) carbodithioates. J. Therm. Anal. Calorim. 2015, 119 (3), 1619-1632.