n-Dodecyltrimethoxysilane, 95%
n-Dodecyltrimethoxysilane, 95%
n-Dodecyltrimethoxysilane, 95%
Thermo Scientific Chemicals

n-Dodecyltrimethoxysilane, 95%

CAS: 3069-21-4 | C15H34O3Si | 290.52 g/mol
Quantity:
25 g
100 g
Catalog number L16486.22
also known as L16486-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS3069-21-4
IUPAC Namedodecyltrimethoxysilane
Molecular FormulaC15H34O3Si
InChI KeySCPWMSBAGXEGPW-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC[Si](OC)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.4235-1.4310 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only