n-Hexyltriethoxysilane, 97%
n-Hexyltriethoxysilane, 97%
n-Hexyltriethoxysilane, 97%
Thermo Scientific Chemicals

n-Hexyltriethoxysilane, 97%

CAS: 18166-37-5 | C12H28O3Si | 248.44 g/mol
Quantity:
25 g
100 g
Catalog number L16521.22
also known as L16521-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS18166-37-5
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4070-1.4120 @ 20°C (non-U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Assay (GC)≥96.0%
FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only