Triethyl 2-fluoro-2-phosphonoacetate, 96%
Triethyl 2-fluoro-2-phosphonoacetate, 96%
Triethyl 2-fluoro-2-phosphonoacetate, 96%
Thermo Scientific Chemicals

Triethyl 2-fluoro-2-phosphonoacetate, 96%

CAS: 2356-16-3 | C8H16FO5P | 242.183 g/mol
Quantity:
1 g
5 g
25 g
Catalog number L17913.06
also known as L17913-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2356-16-3
IUPAC Nameethyl 2-(diethoxyphosphoryl)-2-fluoroacetate
Molecular FormulaC8H16FO5P
InChI KeyFVPISMANESAJQZ-UHFFFAOYNA-N
SMILESCCOC(=O)C(F)P(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Refractive Index1.4225-1.4285 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only