Thermo Scientific Chemicals

Isoprene monoxide, 95%, Thermo Scientific Chemicals

Catalog number: L19768.14
25 g, Each
Thermo Scientific Chemicals

Isoprene monoxide, 95%, Thermo Scientific Chemicals

Catalog number: L19768.14
25 g, Each
Quantity
Catalog number: L19768.14
also known as L19768-14
Price (USD)
Price: 619.00
Online price: 526.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1838-94-4
IUPAC Name
2-ethenyl-2-methyloxirane
Molecular Formula
C5H8O
InChI Key
FVCDMHWSPLRYAB-UHFFFAOYNA-N
SMILES
CC1(CO1)C=C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Refractive Index
1.4145-1.4195 @ 20?C

Description

Isoprene monoxide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoprene monoxide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Recommended storage temperature is 2 - 8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text