2-Fluoro-4-nitrobenzoic acid, 98%
2-Fluoro-4-nitrobenzoic acid, 98%
2-Fluoro-4-nitrobenzoic acid, 98%
Thermo Scientific Chemicals

2-Fluoro-4-nitrobenzoic acid, 98%

CAS: 403-24-7 | C7H4FNO4 | 185.11 g/mol
Quantity:
1 g
5 g
Catalog number L19770.03
also known as L19770-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS403-24-7
IUPAC Name2-fluoro-4-nitrobenzoate
Molecular FormulaC7H3FNO4
InChI KeyMMWFMFZFCKADEL-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=C(C=C1F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)169.0-176.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only