3-Amino-3-phenylpropionic acid, 99%
3-Amino-3-phenylpropionic acid, 99%
3-Amino-3-phenylpropionic acid, 99%
3-Amino-3-phenylpropionic acid, 99%
Thermo Scientific Chemicals

3-Amino-3-phenylpropionic acid, 99%

CAS: 614-19-7 | C9H11NO2 | 165.19 g/mol
Quantity:
1 g
5 g
Catalog number L19801.03
also known as L19801-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS614-19-7
IUPAC Name3-amino-3-phenylpropanoic acid
Molecular FormulaC9H11NO2
InChI KeyUJOYFRCOTPUKAK-UHFFFAOYNA-N
SMILESNC(CC(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
Assay (HPLC)≥98.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only