4-Benzylaniline, 98%
4-Benzylaniline, 98%
4-Benzylaniline, 98%
Thermo Scientific Chemicals

4-Benzylaniline, 98%

CAS: 1135-12-2 | C13H13N | 183.254 g/mol
Quantity:
1 g
5 g
Catalog number L19873.06
also known as L19873-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1135-12-2
IUPAC Name4-benzylaniline
Molecular FormulaC13H13N
InChI KeyWDTRNCFZFQIWLM-UHFFFAOYSA-N
SMILESNC1=CC=C(CC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5% (Non-U.S. Specification)
CommentPurchased in the U.S. and in other countries
FormCrystals or powder or crystalline powder and/or chunks or fused solid
Assay from Supplier's CofA≥97.5% (GC, U.S. Specification)
Melting Point (clear melt)34-44°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only