2-Chloro-3-fluoropyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Chloro-3-fluoropyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals
2-Chloro-3-fluoropyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-3-fluoropyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Quantity:
250 mg
1 g
Catalog number L20024.03
also known as L20024-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS628691-93-0
IUPAC Name2-chloro-3-fluoropyridine-4-carboxylic acid
Molecular FormulaC6H3ClFNO2
InChI KeyNWNWBLRKHOVSEL-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)C(Cl)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Assay (HPLC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only