(S)-(+)-1-(1-Naphthyl)ethyl isocyanate, 95%
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate, 95%
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate, 95%
Thermo Scientific Chemicals

(S)-(+)-1-(1-Naphthyl)ethyl isocyanate, 95%

CAS: 73671-79-1 | C13H11NO | 197.24 g/mol
Quantity:
250 mg
1 g
Catalog number L20242.03
also known as L20242-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS73671-79-1
SpecificationsSpecification SheetSpecification Sheet
CommentMay form a fine precipitate on standing
Refractive Index1.6010-1.6070 @ 20?C
Assay (GC)≥94.0%
Appearance (Color)Clear to hazy, colorless to orange or brown
Optical Rotation+44? ±2? (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only