Naphthalene-1,4-dicarboxylic acid, 98+%
Naphthalene-1,4-dicarboxylic acid, 98+%
Naphthalene-1,4-dicarboxylic acid, 98+%
Thermo Scientific Chemicals

Naphthalene-1,4-dicarboxylic acid, 98+%

CAS: 605-70-9 | C12H8O4 | 216.19 g/mol
Catalog NumberQuantity
L20249.22
also known as L20249-22
100 g
Catalog number L20249.22
also known as L20249-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS605-70-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Tapas Kumar Maji; Wakako Kaneko; Masaaki Ohba; and Susumu Kitagawa. Diversity in magnetic properties of 3D isomorphous networks of Co(II) and Mn(II) constructed by napthalene-1,4-dicarboxylate. Chem. Commun. 2005, (36), 4613-4615.
  2. J. F. Walker; N. D. Scott. Sodium Naphthalene. II. Preparation and Properties of Dihydronaphthalene Dicarboxylic Acids. J. Am. Chem. Soc. 1938, (60), 951-955.