Thermo Scientific Chemicals

L-(-)-Prolinamide, 98%, Thermo Scientific Chemicals

Catalog number: L20417.06
5 g, Each
Thermo Scientific Chemicals

L-(-)-Prolinamide, 98%, Thermo Scientific Chemicals

Catalog number: L20417.06
5 g, Each
Quantity
Catalog number: L20417.06
also known as L20417-06
Price (TWD)

Chemical Identifiers

CAS
7531-52-4
Optical Rotation
-105? ± 3? (c=2 in Ethanol)
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
95-104?C
Appearance (Color)
White to pale yellow
Assay (GC)
≥97.5%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.

Solubility
Soluble in water and ethanol (50 mg/ml).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text