Methanol, Certified AR for Analysis, Fisher Chemical™
Methanol, Certified AR for Analysis, Fisher Chemical™
Methanol, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Methanol, Certified AR for Analysis, Fisher Chemical™

CH4O, CAS Number-67-56-1, wood naphtha, colonial spirit, pyroxylic spirit, methyl hydroxide, methyl alcohol, wood spirit, wood alcohol, methylol, columbian spirit, carbinol, 2.5L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042 | CAS: 67-56-1 | CH4O | 32.04 g/mol
Quantity:
2.5 L
5 L
Packaging:
View all
Plastic-coated Glass Bottle
Amber Glass Bottle
HDPE Bottle
Catalog number M-4000-PB17
also known as M/4000/PB17
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Specifications
Boiling Point64.7°C
CAS67-56-1
ChEBICHEBI:17790
ColorColorless
Formula Weight32.04g/mol
View more
CH4O, CAS Number-67-56-1, methyl alcohol, colonial spirit, wood naphtha, wood spirit, wood alcohol, methylol, pyroxylic spirit, methyl hydroxide, columbian spirit, carbinol, 2.5L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042