Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody recognizes alpha, beta-tubulin heterodimer, a basic intracellular structural unit of microtubules. Alpha- and beta-tubulins form approximately 100 kDa tubulin heterodimer, a globular protein that polymerizes to form microtubules.
Western Blot: Reducing conditions.
Tubulin protein is the major constituent of microtubules. Tubulin is a dimer of alpha and beta chains, which binds two molecules of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain. There are at least six isotypes of both alpha- and beta- tubulin in human cells, which are distinguished by slightly different amino acid sequences and encoded by a large, multigene family that has been highly conserved throughout evolution. Although the most important functions of microtubules in proliferative cells are through their actions as components of the mitotic spindle, they are also involved in many other essential functions throughout the cell cycle of both malignant and nonmalignant cells. Anti-microtubule agents including Vinca alkaloids and taxanes may disrupt many of these essential functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-tubulin; Alpha-tubulin 1; alpha-tubulin isotype M-alpha-1; alpha-tubulin isotype M-alpha-6; beta 5-tubulin; beta Ib tubulin; epididymis secretory sperm binding protein; FLJ30169; H2 ALPHA; hum-a-tub1; hum-a-tub2; Talpha1 alpha-tubulin; Tubulin alpha; tubulin alpha 3; Tubulin alpha-1 chain; tubulin alpha-3 chain; tubulin B-alpha-1; tubulin beta; tubulin beta-1 chain; tubulin beta-5 chain; tubulin, alpha 1; tubulin, alpha, brain-specific; tubulin, beta 1; tubulin, beta 2A class IIa; tubulin, beta family 1; tubulin, beta polypeptide; unnamed protein product; XTP3TPATP1; α/β-tubulin
Gene Aliases: 2810484G07Rik; B-ALPHA-1; CDCBM6; CSCSC1; LIS3; M(beta)1; M40; MACTHC1; OK/SW-cl.56; Tuba-1; Tuba1; TUBA3; TUBB1; TUBB2A; TUBB5
UniProt ID: (Human) Q71U36, (Human) P07437, (Pig) Q767L7, (Human) Q9H4B7, (Mouse) P68369, (Rat) P68370, (Mouse) A2AQ07
Entrez Gene ID: (Pig) 100135051, (Human) 7846, (Human) 203068, (Pig) 733686, (Pig) 100134983, (Human) 81027, (Mouse) 22142, (Rat) 64158, (Rat) 679312, (Mouse) 545486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support