Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This is a secreted chemokine which attracts macrophages during inflammation and metastasis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues. The protein is an in vivo substrate of matrix metalloproteinase 2, an enzyme which degrades components of the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chemokine (C-C motif) ligand 7; H-MCP-3; MGC138463; MGC138465; monocyte chemoattractant protein 3; monocyte chemotactic protein 3; RP23-350G1.4; small inducible cytokine A7 (monocyte chemotactic protein 3); small-inducible cytokine A7
Gene Aliases: FIC; MARC; MCP-3; MCP3; NC28; SCYA6; SCYA7
UniProt ID: (Human) P80098
Entrez Gene ID: (Human) 6354
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support