Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IGF1R (Insulin-like Growth Factor1 Receptor) is a receptor tyrosine kinase that mediates actions of insulin-like growth factor 1 (IFG1). It binds IGF1 with high affinity and IGF2/insulin with lower affinity. Activated IGF1R is involved in cell growth and survival control. IGF1R is crucial for tumor transformation and survial of malignant cells. Ligand binding activates the receptor kinase and leads to receptor autophosphorylation and tyrosines phosphorylation of multiple substrates that function as signaling adapter proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD221; IGF-I receptor; Insulin-like growth factor 1 receptor; Insulin-like growth factor I receptor
Gene Aliases: CD221; IGF1R; IGFIR; IGFR; JTK13
UniProt ID: (Human) P08069
Entrez Gene ID: (Human) 3480
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support