Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: 293T, A431, HeLa, HepG2, human SLU7-transfected 293T.
Predicted reactivity: Human (99%), Mouse (98%), Rat (97%), Zebrafish (80%), Chicken (86%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC9280; similar to Saccharomyces cerevisiae splicing factor Slu7; SLU7 homolog, splicing factor; SLU7 splicing factor homolog; splicing factor; step II splicing factor SLU7; unnamed protein product; zinc knuckle motif containing
Gene Aliases: 9G8; hSlu7
UniProt ID: (Human) O95391
Entrez Gene ID: (Human) 10569
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support