Search
Search
FIGURE: 1 / 5
Positive Samples: A-549, Mouse lung, Rat brain
Immunogen sequence: QTIVKLGNKV QELEKQQVTI LATPLPEESM KQELQNLRDE IKQLGREIRL QLKAIEPQKE EADENYNSVN TRMRKTQHGV LSQQFVELIN KCNSMQSEYR E
STX4 is a plasma membrane t-SNARE that mediates docking of transport vesicles.STX4 is necessary for the translocation of SLC2A4 from intracellular vesicles to the plasma membrane. Together with STXB3 and VAMP2, STX4 may also play a role in docking/fusion of intracellular GLUT4-containing vesicles with the cell surface in adipocytes. STX4 may also play a role in docking of synaptic vesicles at presynaptic active zones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Renal carcinoma antigen NY-REN-31; STX4A; syntaxin 4A (placental); Syntaxin-4; unnamed protein product
Gene Aliases: DFNB123; p35-2; STX4; STX4A; Syn-4; Syn4
UniProt ID: (Human) Q12846, (Rat) Q08850, (Mouse) P70452
Entrez Gene ID: (Human) 6810, (Rat) 81803, (Mouse) 20909
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support