InSight Mote Transmitters
Actual product may vary
Thermo Scientific™
InSight Mote Transmitters
Thermo Scientific™ InSight mote transmitters periodically wake up and transmit sensor data to the InSight link server.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호채널
MOTET3C1D2A111108
MOTET3C1D2A111137
카탈로그 번호 MOTET3C1D2A11110
제품 가격(KRW)
-
채널:
8
Thermo Scientific™ InSight battery operated motes are configured during installation for your wireless network. Periodically, they wake up and transmit sensor data to the InSight link server. In the case of communication failure, the mote will log data for up to 24 hours.
  • Motes are available for WiFi and 915MHz wireless servers (North America)
  • Warranty varies by country, please contact us for details
Not available outside North America
사양
배터리 종류C
보정Not Calibrated
채널8
설명InSight mote, 915MHz transmitter, 8-channels: 3-temp, 1-KVA/50 amps, ext. power connector, 2-door with “C” battery & enclosure. Sensors not included.
디스플레이 유형No Display
유형Insight Remote
Unit SizeEach