Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
Fisher Chemical™

Nitric Acid 68% d=1.42, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™

HNO3, CAS Number-7697-37-2, hydrogen nitrate, nital, aqua fortis, azotic acid, acide nitrique, acidum nitricum, rfna, salpetersaeure, nitrous fumes, nitryl hydroxide, 2.5L, CHEBI:48107, Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, HDPE plastic bottle | CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Quantity:
1 L
2.5 L
Catalog number N-2272-PB15
also known as N/2272/PB15
Price (JPY)
-
Quantity:
1 L
Request bulk or custom format
Specifications
CAS7697-37-2
ChEBICHEBI:48107
Chemical Name or MaterialNitric Acid
ColorYellow
Formula Weight63.02g/mol
View more
HNO3, CAS Number-7697-37-2, hydrogen nitrate, nital, aqua fortis, azotic acid, acide nitrique, acidum nitricum, rfna, salpetersaeure, nitrous fumes, nitryl hydroxide, 2.5L, CHEBI:48107, Yellow, 63.02g/mol, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -41 deg.C, 11349, 63.012, HDPE plastic bottle