NanoDrop™ OneC Microvolume UV Spectrophotometer - FAQs
NanoDrop™ OneC Microvolume UV Spectrophotometer - FAQs
View additional product information for NanoDrop™ One/OneC Microvolume UV-Vis Spectrophotometer - FAQs (ND-ONE-W, ND-ONEC-W)
51 product FAQs found
Which is the best DNA or RNA quantification instrument?
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
After servicing NanoDrop instruments, do you provide a service report listing the repairs made to the instrument?
A report is shipped back with each NanoDrop instrument listing the services that were performed and any parts that were replaced. We also provide you with an Instrument Certification Sheet which shows that the instrument passed recalibration and recertification.
Do you provide “as found” and “after service” data for servicing of NanoDrop instruments?
If you specifically request “as found” and “after service” data when setting up the NanoDrop instrument service, we can provide results of a Performance Verification before and after the servicing.
Will you send me a shipping label or can I use my own shipping provider to send in a NanoDrop instrument for servicing?
Upon receipt of confirmation of payment method, we will provide you a FedEx return shipping label and shipping instructions. Alternatively, we will waive the shipping cost if you decide to use your own carrier.
Do I need a special shipping box for my NanoDrop instrument to send for servicing? If so, would you supply the shipping box and what is the cost?
The NanoDrop 2000/2000c, NanoDrop 8000, NanoDrop Eight, NanoDrop Lite, NanoDrop Lite Plus, and NanoDrop One/OneC instruments all need to be shipped in their original shipping box and packing foam. If you do not have those materials, we will send you a box and packing foam at no cost. The NanoDrop 3300 instrument can be shipped in any box you may have available.
Does the NanoDrop instrument service cost include part replacements?
The cost for NanoDrop instrument service does not include any parts. If a part replacement is needed, we will contact you to obtain your authorization to replace the part.
How do I pay for service for my NanoDrop instrument?
You can pay for NanoDrop instrument service with a purchase order or credit card. We accept Master Card, Visa, or American Express.
What is the turnaround time for servicing NanoDrop instruments in the United States and Canada?
The turnaround time for servicing NanoDrop instruments in the United States is 48 to 72 hours. The turnaround time in Canada is approximately 2 days longer, depending upon the time spent in Customs in either direction. FedEx weather-related delays can also extend the turnaround time.
Do you have onsite technicians for servicing NanoDrop instruments in the United States and Canada?
We do not have onsite technicians to perform service for Thermo Scientific NanoDrop instruments located in the United States and Canada. All service for Thermo Scientific NanoDrop instruments is performed by our trained service technicians at our service depot in Madison, WI.
Whom do I contact to initiate service for my NanoDrop instrument?
For NanoDrop instrument service in the United States and Canada, call 877-724-7690 (toll-free) and select option 4 for Technical Support or email nanodrop@thermofisher.com. For international service, please contact your local NanoDrop distributor.
Can I perform my own NanoDrop instrument service?
All service for Thermo Scientific NanoDrop instruments is performed by our trained service technicians at our service depot in Madison, WI.
Which protein assays can I perform using the NanoDrop 2000, NanoDrop One, NanoDrop 8000, NanoDrop Eight, NanoDrop Lite, and NanoDrop Light Plus spectrophotometers?
Visit this website (https://www.thermofisher.com/us/en/home/industrial/spectroscopy-elemental-isotope-analysis/molecular-spectroscopy/ultraviolet-visible-visible-spectrophotometry-uv-vis-vis/uv-vis-vis-instruments/nanodrop-microvolume-spectrophotometers/nanodrop-protein-quantification.html) for useful guidance on protein assays for NanoDrop One, NanoDrop Eight, and NanoDrop Lite spectrophotometers. View this guide (https://tools.thermofisher.com/content/sfs/manuals/T032-NanoDrop-2000-2000c-Protein-Measurements.pdf) for protein assays for NanoDrop 2000 spectrophotometer. View this guide https://tools.thermofisher.com/content/sfs/brochures/T010-ND-1000-&-ND-8000-Protein-Measurements.pdf) for NanoDrop 8000 spectrophotometer. Colorimetric assays such as the Pierce 660 nm, BCA, Bradford and Lowry, are generally used for uncharacterized protein solutions and cell lysates. Proteins that contain Trp, Tyr residues, or Cys-Cys disulphide bonds will absorb in the UV range (i.e., 280 nm) making absorbance spectroscopy a fast, convenient method for the quantitation of purified protein preparations using the Protein A280 application module.
What is the Beer-Lambert equation?
Beer-Lambert Equation is as follows:
A = E * b * c
- A is the absorbance of the sample
- E is the wavelength-dependent molar absorptivity coefficient (or extinction coefficient) expressed in units of L/mol-cm
- b is the pathlength in cm
- c is the analyte concentration in mol/L or molarity (M).
For Nucleic Acid calculations, the Beer-Lambert equation is modified to use an extinction coefficient with units of ng-cm/mL. Using this extinction coefficient gives a manipulated equation:
c = (A * e)/b
- c is the nucleic acid concentration in ng/µL
- A is the absorbance of the sample
- e is the nucleic acid specific concentration factor in ng-cm/µL (dsDNA = 50 ng/µL; ssDNA = 33 ng/µL; RNA = 40 ng/µL; dsRNA = 40 ng/µL)
- b is the pathlength in cm
When using a NanoDrop spectrophotometer, what does 10 mm equivalent absorbance mean?
NanoDrop spectrophotometers use light pathlengths from 1.0 mm to as low as 0.05 mm (model dependent) to make measurements, allowing the quantification of samples up to 50 or 200 times more concentrated than traditional cuvette based spectrophotometers are able to measure. Applications such as Nucleic Acid and Protein A280 will display this absorbance as a 10 mm equivalent absorbance.
Are the sample concentrations I obtain with the NanoDrop spectrophotometer pedestal volume dependent?
The sample concentrations are calculated according to Beer's Law which is volume independent. Beer's Law relates absorbance to concentration using a wavelength specific molar extinction coefficient or the use of a nucleic acid concentration factor and a pathlength of 1 cm.
Which pathlength was used for my sample measurement on the NanoDrop spectrophotometer pedestal?
The software is programmed to determine the best pathlength for the measurement based upon the absorbance signal of the sample at the analysis wavelength for the Nucleic Acids module, Protein A280 module, and Custom methods. On the other hand, the autopathlength function in the UV-Vis module monitors the entire wavelength range to optimize pathlength selection. The absorbance signal is normalized to a either 10 mm or 1.0 mm pathlength equivalent depending on module.
When using a NanoDrop spectrophotometer, how does data collected in UV-Vis module compare to the data collected in another application?
UV-Vis data is reported at a normalized 1.0 mm pathlength. As a result, absorbance values reported here will be reported 10-fold lower than those reported by the Nucleic Acid or Protein A280 applications.
When using a NanoDrop spectrophotometer, when do I use the autopathlength function in the UV-Vis module?
Selecting autopathlength should be considered if the peak of interest is greater than 1.25 A at a 1.0 mm pathlength or to ensure that the entire sample absorbance spectrum remains within the linear response of the detector. With autopathlength deselected, the instrument uses only a 1.0 mm pathlength which is optimized for absorbance signal less than 1.25 A.
When using a NanoDrop spectrophotometer, what are the advantages of the Pierce 660 assay over the Bradford assay?
Both assays are mix-and-measure (minimal incubation times and no heating steps). Pierce 660 Assay has a more stable end-point. Pierce 660 reaction will not continue to progress and form aggregates.
When performing a Pierce 660 assay with a NanoDrop spectrophotometer, why am I experiencing lower absorbance on the NanoDrop pedestal compared to the cuvette?
When utilizing the measurement pedestal, NanoDrop spectrophotometers will report the Pierce 660 assay absorbance using a 1.0 mm pathlength. As a result, the absorbance displayed will be 10-fold lower than if measured in a conventional 10 mm cuvette. This reduction in signal does not compromise the sensitivity or linear range of the assay.
When performing a Lowry protein assay, why am I experiencing low absorbance signal across the dynamic range of the assay when using a NanoDrop spectrophotometer?
The Lowry module reports absorbance signal at a 1.0 mm pathlength rather than the conventional 10 mm pathlength resulting in a 10-fold reduction in absorbance signal. Consequently, the reduction in signal will not compromise the sensitivity or dynamic range of the assay. In the cases of extreme low signal, it is typically due to the formulation of the Lowry reagent from the specific vendors. We have had the greatest success with the Pierce product line of colorimetric assays.
General Information: Wavelength (modified Lowry protocol for a NanoDrop spectrophotometer) is measured at 650 nm and normalized at 405 nm Dynamic range of assay is 0.2 mg/mL to 4.0 mg/mL (BSA)
How can insufficient mixing affect my Bradford assay when using a NanoDrop spectrophotometer?
A poorly mixed Bradford assay will not properly complete and may form a gradient in the tube. This can affect reproducibility.
Why am I experiencing low absorbance formulations, when performing Bradford assays with my NanoDrop spectrophotometer?
Some Bradford assay manufacturers' formulations will not provide sufficient absorbance to obtain an accurate and reproducible standard curve.
Why is the reproducibility of my Bradford assay outside the manufacturer's specification with a NanoDrop spectrophotometer?
The Bradford reagent will form particulates as a reaction progresses. These particulates may cause light scatter during a sample measurement, resulting in higher than expected variation between measurements. Particulates can be minimized by limiting the incubation time to 10 min prior to measuring as per the Thermo Scientific Pierce protocol. In addition, a sample volume of 2 µL should be used for all colorimetric assay measurements.
What is the importance of uniform sample heating when performing a BCA assay with a NanoDrop spectrophotometer?
To ensure accurate results, sample heating must be uniform for both standards and unknown samples.
When performing a BCA assay with a NanoDrop spectrophotometer, why am I obtaining a lower absorbance signal on the NanoDrop spectrophotometer pedestal compared to a conventional cuvette system?
When utilizing the measurement pedestal, NanoDrop spectrophotometers will report the BCA assay absorbance using a 1.0 mm pathlength. As a result, the absorbance displayed will be 10-fold lower than if measured in a conventional 10 mm cuvette. This reduction in signal does not compromise the sensitivity or linear range of the assay.
When I am using a NanoDrop spectrophotometer, what is the appropriate ratio for a pure protein sample?
A purified protein should show a 260/280 purity ratio of approximately 0.63.
Why is it not recommended that I use RIPA buffer for protein A280 measurements with my NanoDrop spectrophotometer?
RIPA buffer produces a particularly strong absorbance signal at the 280 nm wavelength. As a result, it will either over estimate or under estimate protein concentrations and interfere with the protein purity ratio.
Protein samples in RIPA buffer should be quantified via the Pierce Protein 660 or BCA colorimetric assays using a full spectrum NanoDrop model.
Find additional tips, troubleshooting help, and resources within ourProtein Purification and Isolation Support Center.
When I am using a NanoDrop spectrophotometer to calculate protein concentrations, what is the difference between E1% and E0.1% value?
The difference between the E1% and E0.1% values are the associated units for the results.
The E1% value has units of a 1% solution or 1 g/100mL. In the NanoDrop software this value is converted to an E0.1% value to display protein concentrations in mg/mL units.
Please refer to Tech Tip #6: Extinction Coefficients for further detail: https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0006-Extinction-coefficients.pdf
How are protein concentrations calculated by the Protein A280 module in the NanoDrop spectrophotometer?
Protein concentrations obtained from the Protein A280 module are calculated using a 1% solution extinction coefficient (E1%) in place of the molar extinction coefficient. Each purified protein will have a unique E1% value which can be obtained from either references or, if produced commercially, from the vendor. The user may choose to use a known E1% value or input the molar extinction coefficient and molecular weight of the protein into the NanoDrop software to obtain the mg/mL protein concentrations.
If only a rough estimation is desired, the 1 Abs = 1 mg/mL can be used instead of a specific E1% value.
Does my NanoDrop One/Onec Spectrophotometer require a computer to operate?
No. These instruments feature an on-board, high resolution touchscreen control with an Android-based operating system and 32 GB flash memory. USB, Ethernet, and Wi-Fi connectivity options allow seamless data transfer to an external computer with Windows 7 or 10, 64-bit operating systems. The instrument can be controlled using the onboard touchscreen or a computer that is connected to the instrument. Customers can download the NanoDrop One PC software from our website to operate the system, view and analyze their data, and explore Acclaro support features from their desks.
Can I verify that my NanoDrop One/Onec Spectrophotometer is calibrated and running accurately?
Yes. While most users will never need to recalibrate their NanoDrop One/Onec Spectrophotometer, we recommend verifying instrument performance using the NanoDrop PV-1 Performance Verification Solution every 6 months. PV-1 is available from the Thermo Fisher website.
Is simply wiping the pedestal surfaces of my NanoDrop One/Onec Spectrophotometer enough to prevent carryover?
Yes. The highly polished quartz and stainless steel surfaces of the sample retention system are resistant to sample adherence, making the use of dry lab wipes very effective in removing the sample.
What level of measurement performance should I expect with my NanoDrop One/Onec Spectrophotometer?
The photometric accuracy is within 3% at 0.97 A at 302 nm. Typical measurement reproducibility is 0.002 A (1.0 mm path) or 1% CV, whichever is greater.
Can I measure purified and unpurified proteins with my NanoDrop One/Onec Spectrophotometer?
Purified proteins can be measured directly using the A280 or A205 pre-configured applications. Choose the most suitable extinction coefficient from the pre-defined protein sample types (BSA, IgG, Lysozyme) from our drop-down list or add your own custom protein using the Protein Editor feature.
For unpurified proteins or cell lysates, a selection of pre-configured colorimetric assay applications is available.
Is there a feature, on my NanoDrop One/Onec Spectrophotometer, that allows me to create and save custom methods?
Yes, within our NanoDrop One/Onec spectrophotometer, methods can be built with customized analysis wavelengths, factors, extinction coefficients and more. Fluorescent labels can be saved in the Dye Chromophore editor and new protein methods can be saved in the Protein Editor.
What pre-configured applications are included in my NanoDrop One and NanoDrop Onec spectrophotometers?
Listed below are the pre-configured applications included in our NanoDrop One/Onec Spectrophotometers:
- Nucleic Acid A260 (ds DNA, ssDNA, oligo DNA, oligo RNA, custom factor), A260/280, A260/230 and Microarray (labeled nucleic acids)
- Protein A280 and A205, Protein Pierce 660, Protein Bradford, Protein BCA, Protein Lowry, Proteins and labels
- OD600
- Kinetics, UV-Vis and Custom Methods
What is the difference between your NanoDrop One Spectrophotometer and your NanoDrop Onec Spectrophotometer?
Both models have the patented microvolume sample retention capability (pedestal).
However, the NanoDrop Onec Spectrophotometer has a built-in cuvette capability that allows for all the same pedestal applications plus cuvette measurements to support kinetics applications and dilute sample measurements. The cuvette position can be used with the arm up or down and comes with temperature control and stirring features.
How does Acclaro Contaminant Identification feature help me troubleshoot my NanoDrop spectrophotometer?
When the user taps the information icon on the measurement screen, a popup box will report the sample issue detected and offer possible causes and solutions. If the user wants to "Learn More" they will be guided through a cascade of information, from fundamentals to the specifics.
This is best explained with an example:
When Acclaro detects a low A260/A280 result, a popup box will inform the user that the ratio is outside acceptable limits for pure DNA sample and that this could be due to a poor blank or a contaminant. The user will then see information on common contaminants that can affect this ratio, and other resources starting with an animation that explains "What is a purity ratio." If protein is the more likely contaminant, the user can look at spectra of DNA/protein samples and how increasing protein levels can affect the purity ratios of a DNA sample.
Which contaminants can I detect using the Acclaro Contaminant Identification feature?
The Acclaro Contaminant Identification feature currently supports detection of the following possible contaminants:
- In dsDNA samples: proteins, phenol, guanidine HCl, and guanidine isothiocyanate
- In RNA samples: proteins, phenol, and guanidine isothiocyanate
- In protein samples (direct A280 measurements): nucleic acids and phenol
What is the Acclaro Sample Intelligence technology?
Acclaro Sample Intelligence technology enhances user understanding of sample quality while delivering accurate quantitative measurements. There are three parts to Acclaro:
- data analysis algorithms that provide contaminant identification and corrected concentrations
- an embedded sensor and digital image analysis that monitors the sample column for bubbles to ensure measurement integrity
- sample information alerts and on-demand technical support for guided troubleshooting
How are the NanoDrop One/Onec spectrophotometers different from traditional cuvette-based instruments?
NanoDrop One/Onec Spectrophotometers use a patented microvolume sample retention system to minimize sample consumption and eliminate the need for cuvettes. A variable, auto-range pathlength feature allows users to measure up to 366x higher sample concentrations than can be measured in a 10 mm cuvette making dilution steps unnecessary.
Do I need to recondition the pedestals of my NanoDrop spectrophotometer?
Pedestal surfaces may become "un-conditioned" and possibly inhibit proper liquid column formation required for measurement precision. Refer to this Cleaning and Reconditioning document for more information: http://tools.thermofisher.com/content/sfs/brochures/T005-NanoDrop%201000-&-NanoDrop%208000-Cleaning-and-Reconditioning.pdf
What if I use my NanoDrop spectrometer to measure a sample that is too diluted?
Measuring at or below the detection limit may result in SD values outside the precision specification. Refer to instrument user manual for detection limits for specific applications.
Can I make multiple measurements on a single aliquot when using a NanoDrop spectrophotometer?
Evaporation can occur when samples remain on the pedestal beyond the time required to make one measurement, therefore we recommend always using a fresh aliquot for each measurement.
Why am I experiencing poor sample homogeneity when using my NanoDrop spectrophotometer?
Poor sample homogeneity is commonly associated with non-homogeneous DNA samples. Due to the small sample volumes used for pedestal measurements it is important to mix the sample thoroughly. Samples containing large molecules such as genomic or lambda DNA are particularly susceptible to this phenomenon. If compatible with the protocol being used, heat the DNA samples to 63 degrees C and lightly finger vortex before measurement to ensure the nucleic acid is properly in solution.
When using a NanoDrop spectrophotometer, what is the difference between controls versus standards?
NanoDrop CF-1 Calibration Fluid is the only photometric standard available to assess the accuracy of the NanoDrop spectrophotometer pedestal. If the control sample shows unexpected results the user should confirm the calibration of the instrument with the CF-1 fluid.
Some control examples include:
- Nucleic Acids: Fermentas GeneRuler Express DNA Ladder 100-5000 bp (Cat. No. SM1551)
- Protein: Pierce Bovine Serum Albumin Standard Ampules, 2 mg/mL (Cat. No. 23209)
How can I check the accuracy of my NanoDrop spectrophotometer?
The accuracy of NanoDrop spectrophotometers can be assessed by using NanoDrop CF-1 Calibration Fluid in conjunction with the calibration check application in the NanoDrop software.
What are acceptable purity ratios, when using a NanoDrop spectrophotometer?
Please review the following information regarding acceptable purity ratios, when using NanoDrop spectrophotometers:
- Nucleic acid samples: A260/A280 ratios of 1.8-2.2; A260/A230 ratios of 1.8-2.2 are generally considered pure
- Protein samples: A260/A280 ratio of approximately 0.63 is generally considered pure
When using a NanoDrop spectrophotometer, what would cause my 260/280 purity ratios to fall outside of the generally accepted range?
Please review the following possible causes for 260/280 purity ratios to fall outside of the generally accepted range when using a NanoDrop spectrophotometer:
- The starting sample or the extraction method may result in contaminants in the final extract, e.g. guanidine (often from column based kits), phenol carryover or carbohydrates from the original cells (often a problem with plant samples). This is often best diagnosed by looking at the spectra, as many contaminants have characteristic profiles.
- Improper Blank: An incorrect or highly absorbent blanking solution or sample residue on the measurement pedestal may affect purity ratios.
- Sample Concentration: If the sample is nearing the lower detection limit of the instrument. Contributions from instrument noise may affect the shape of the spectrum. Check instrument specifications for lower detection limits. Purity ratios cannot be accurately calculated for samples close to the lower detection limits of the instrument. If a NanoDrop 2000c is being used, a cuvette may be used for samples at very low concentrations.