MAS™ Omni·CARDIO™ Control
Actual product may vary
Thermo Scientific™
MAS™ Omni·CARDIO™ Control
Ensure accurate results in testing of traditional cardiac marker panels and the new generation of STAT analytes including D-Dimer, hCG, Myeloperoxidase and Procalcitonin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
OCRD-2022
OCRD-ULUltra Low
OCRD-LLow
OCRD-1011
OCRD-3033
OCRD-MPTri-Level Multi-Pack
Catalog number OCRD-202
Price (USD)
246.00
Each
Level:
2
Price (USD)
246.00
Each
Ensure accurate results in testing of traditional cardiac marker panels as well as the new generation of STAT analytes including D-Dimer, hCG, Myeloperoxidase and Procalcitonin with the Thermo Scientific™ MAS™ Omni·CARDIO™ Control.
  • Matrix: Human Serum
  • Packaging: 6 x 3mL assayed
  • Value assignment provided for key instrument systems
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Level2
Packaging6
Quantity3 mL
TypeControl
Detectable Analytesβ-hCG, Brain Natriuretic Peptide-32 (BNP-32), Creatinine Kinase-MB (CK-MB), D-dimer, Digitoxin, High Sensitivity C-Reactive Protein (hsCRP), hCG, Myeloperoxidase (MPO), Myoglobin, N-Terminal Brain Natriuretic Peptide (NT-proBNP), Procalcitonin (PCT), Total Creatinine Kinase (Total CK), Troponin I, Troponin T
Sample TypeSerum
Unit SizeEach
Contents & Storage
  • 36 month shelf life from date of manufacture when stored at -25°C to -15°C
  • 15 days at 2°C to 8°C (for BNP-32, CK-MB, D-Dimer, Digitoxin, hCG, hsCRP, Myeloperoxidase, Procalcitonin, Total CK, Troponin-I and Troponin-T)
  • 10 days at 2°C to 8°C (for Myoglobin and NT-proBNP)