Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NFAT5 Antibodies

          Invitrogen

          NFAT5 Polyclonal Antibody

          Advanced Verification
          32 Published Figures
          37 References
          View all (14) NFAT5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NFAT5 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          • Published Figures (32)
          • Advanced Verification (6)
          NFAT5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NFAT5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 46

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NFAT5 Antibody (PA1-023) in ICC/IF

          Immunofluorescent analysis of NFAT5 using anti-NFAT5 polyclonal antibody (Product # PA1-023) (shown in green) in HeLa cells. Formalin fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 10 minutes at room temperature. Cells were then blocked with 1% Blocker BSA (Product # 37525) for 15 minutes at room temperature. Cells were probed with a rabbit polyclonal antibody recognizing NFAT5 (Product # PA1-023), at a dilution of 1:100 for at least 1 hour at room temperature. Cells were washed with PBS and incubated with DyLight 488 goa... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NFAT5 Antibody in Immunocytochemistry (ICC/IF)
          NFAT5 Antibody in Immunocytochemistry (ICC/IF)
          NFAT5 Antibody in Immunocytochemistry (ICC/IF)
          NFAT5 Antibody in Immunocytochemistry (ICC/IF)
          NFAT5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NFAT5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Immunoprecipitation (IP)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Immunocytochemistry (ICC/IF)
          NFAT5 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in Western Blot (WB)
          NFAT5 Antibody in ChIP Assay (ChIP)
          NFAT5 Antibody in ChIP Assay (ChIP)
          NFAT5 Antibody
          NFAT5 Antibody
          NFAT5 Antibody
          NFAT5 Antibody
          NFAT5 Antibody
          NFAT5 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NFAT5 Polyclonal Antibody

          Product Details

          PA1-023

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 26 publications 26 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:1,000
          View 4 publications 4 publications

          Immunoprecipitation (IP)

          3 µg
          -

          ChIP assay (ChIP)

          -
          View 2 publications 2 publications

          Gel Shift (EMSA)

          Assay-dependent
          -

          CUT&RUN (CUT&RUN)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse, Rabbit, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues C D(1439) L L V S L Q N Q G N N L T G S F(1455) of human NFAT5.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2152617

          Product Specific Information

          PA1-023 detects nuclear factor of activated T-cells 5 (NFAT5) from human and mouse cells as well as recombinant human protein.

          PA1-023 has been successfully used in Western blot, immunofluorescence, immunoprecipitation, gel shift and immunocytochemistry procedures. By Western blot, this antibody detects an ~170 kDa protein representing NFAT5 in HEK293 cells transfected with the human NFAT5 gene. Immunocytochemical staining of NFAT5 in HEK293 cells transfected with the human NFAT5 gene with PA1-023 results in primarily cytoplasmic staining.

          The PA1-023 immunogen is a synthetic peptide corresponding to residues C D(1439) L L V S L Q N Q G N N L T G S F(1455) of human NFAT5. This sequence is 94% conserved in the mouse protein. PA1-023 immunizing peptide (Cat. # PEP-123) is available for use in neutralization and control experiments.

          Target Information

          The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: glutamine rich protein H65; NFAT; NFAT-like protein 1; NFAT5b; nuclear factor of activated T-cells 5, tonicity-responsive; osmotic response element-binding protein; rel domain-containing transcription factor NFAT5; rel similarity domain containing protein; T-cell transcription factor NFAT5; TonE-binding protein; TonEBP; transcription factor; tonicity-responsive enhancer binding protein; tonicity-responsive enhancer-binding protein; transcription factor; NFAT5; unnamed protein product

          View more View less

          Gene Aliases: B130038B15Rik; CAG-8; CAG80; mKIAA0827; NF-AT5; NFATL1; NFATZ; OREBP; TONEBP

          View more View less

          UniProt ID: (Human) O94916, (Mouse) Q9WV30

          View more View less

          Entrez Gene ID: (Human) 10725, (Mouse) 54446

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding sequence-specific double-stranded DNA binding
          Process(es)
          transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter response to stress cellular response to DNA damage stimulus signal transduction calcineurin-NFAT signaling cascade positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of transcription from RNA polymerase II promoter R-loop processing positive regulation of mitochondrial translation cellular response to cytokine stimulus cellular hyperosmotic response regulation of intracellular signal transduction positive regulation of leukocyte adhesion to vascular endothelial cell renal water homeostasis response to water deprivation positive regulation of gene expression negative regulation of urine volume regulation of calcineurin-NFAT signaling cascade
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch