Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody recognizes a 130 kDa protein from mouse and rat brain tissue extract in western blot.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Neuropilin 1 is a 923 amino acid containing membrane bound coreceptor to a tyrosine kinase receptor belonging to the neuropilin family with two CUB domains, two F5/8 type C domains and a MAM domain. Neuropilin 1 forms a heterodimer with NRP2 (Neuropilin 2) and is known to bind to plexin (PLXNB1) to form semaphorin receptors. Expressed mostly in the nervous system this receptor is implicated in a wide range of functions including in the development of the cardiovascular system, angiogenesis, repulsive axon guidance, formation of certain neuronal circuits, cell survival, control of neuronal migration and organogenesis outside the nervous system. This semaphorin receptor functions as receptors for both the semaphorins and vascular endothelial growth factor and is also a regulator of VEGF-induced angiogenesis. Neuropilin 1 may be involved in aggressive advanced prostate carcinoma and in the progression of breast cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A5 protein; alternatively spliced; lacks exon 16; Neuropilin; Neuropilin-1 precursor (A5 protein); Neuropilin1; NRP 1; receptor for VEGF165 and semaphorins class 3; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; type I transmembrane protein; unnamed protein product; vascular endothelial cell growth factor 165 receptor; VEGF 165 receptor
Gene Aliases: BDCA4; C530029I03; CD304; NP-1; NP1; NPN-1; Npn1; NRP; VEGF165R
UniProt ID: (Human) O14786, (Rat) Q9QWJ9, (Mouse) P97333
Entrez Gene ID: (Human) 8829, (Rat) 246331, (Mouse) 18186
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support