Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 8
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LSAFCLLEAA LAAEVKKPAA AAAPGTAEKL SPKAATLAER SAGLAFSLYQ AMAKDQAVEN ILVSPVVVAS SLGLVSLGGK ATTASQAKAV LSAEQLRDEE VHAGLGELLR SLSNSTARNV TWKL
Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 88%.
SERPINH1 is a member of the serpin superfamily of serine proteinase inhibitors, and its expression is induced by heat shock. The SERPINH1 protein localizes to the endoplasmic reticulum lumen and binds collagen; thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to SERPINH1 have been found in patients with rheumatoid arthritis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 47 kDa heat shock protein; arsenic-transactivated protein 3; AsTP 3; AsTP3; beta enolase repressor factor 1; CBP 1; CBP 2; cell proliferation-inducing gene 14 protein; collagen binding protein 1; Colligen; colligin; Colligin 1; Colligin 2; colligin-1; colligin-2; heat shock protein 47; HGNC 1547; Hsp 47; PIG 14; RA A47; rheumatoid arthritis antigen A-47; rheumatoid arthritis-related antigen RA-A47; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2; serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 2, (collagen-binding protein 2); serine protease inhibitor J6; serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1); unnamed protein product
Gene Aliases: AsTP3; BERF-1; CBP1; CBP2; gp46; HSP47; J6; OI10; PIG14; PPROM; RA-A47; SERPINH2
UniProt ID: (Human) P50454, (Mouse) P19324
Entrez Gene ID: (Human) 871, (Mouse) 12406
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support