Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MLWKLLLRSQ SCRLCSFRKM RSPPKYRPFL ACFTYTTDKQ SSKENTRTVE KLYKCSVDIR KIRRLKGWVL LEDETYVEEI ANILQELGAD ETAVASILER CPEAIVCSPT AVNTQRKLWQ LVCKNEEELI KLIEQFPESF FTIKDQENQK LNVQFFQELG LKNVVISRLL TAAPNVFHNP VEKNKQMVRI LQESYLDVGG SEANMKVWLL KLLSQNPFIL LNSPTAIKET LEFLQEQGFT SFEILQLLSK
Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD3 was initially identified through differential display analysis and is expressed primarily in heart, liver, pancreas and skeletal muscle. MTERFD3 is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription, and may be a serum-inhibitory factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial transcription termination factor-like protein; MTERF domain containing 3; mTERF domain-containing protein 3, mitochondrial; mTERF-like; unnamed protein product
Gene Aliases: 1700007D05Rik; MTERFD3; mTERFL; RGD1311836
UniProt ID: (Human) Q49AM1, (Mouse) Q8BKY8, (Rat) Q5XIE2
Entrez Gene ID: (Human) 80298, (Mouse) 74238, (Rat) 366856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support