Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
This antibody has been tested in direct-ELISA
Methylates arginine residues of myelin basic protein (MBP) in vitro. Methylates symmetrically histone H4 of the FLC chromatin to form H4R3me2s, which in turn suppresses FLC expression to induce flowering. Regulates alternative splicing by methylating spliceosomal proteins. Involved in the post-transcriptional regulation of the circadian clock.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AtPMRT15; Protein arginine N-methyltransferase 1.5; Shk1 kinase-binding protein 1 homolog
Gene Aliases: AT4G31120; ATPRMT5; calcium underaccumulation 1; CAU1; F6E21.40; F6E21_40; PMRT15; PMRT5; PRMT5; PROTEIN ARGININE METHYLTRANSFERASE 5; SHK1 binding protein 1; SKB1
UniProt ID: (Arabidopsis) Q8GWT4
Entrez Gene ID: (Arabidopsis) 829240
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support