TheraPure™ GMP Nucleotides
Thermo Scientific™

TheraPure™ GMP Nucleotides

TheraPure GMP nucleotides help accelerate mRNA vaccine and therapeutics production from preclinical development to commercialization.
Catalog NumberProduct TypeConcentrationQuantityFormat
R00017SKB002UTP, tris solution200 mM5 mLBusiness supply
R00014SKB002ATP, tris solution200 mM5 mLBusiness supply
R00014SKB005ATP, tris solution200 mM50 mLBusiness supply
R00014SKB004ATP, tris solution200 mM500 mLBusiness supply
R044SKB001ATP, sodium solution100 mM3.5 mLBusiness supply
R044SKB002ATP, sodium solution100 mM17 mLBusiness supply
R0441SKB003ATP, sodium solution100 mM50 mLBusiness supply
R0441SKB009ATP, sodium solution100 mM500 mLBusiness supply
R00015SKB002CTP, tris solution200 mM5 mLBusiness supply
R00015SKB005CTP, tris solution200 mM50 mLBusiness supply
R00015SKB004CTP, tris solution200 mM500 mLBusiness supply
R045SKB001CTP, sodium solution100 mM3.5 mLBusiness supply
R045SKB002CTP, sodium solution100 mM17 mLBusiness supply
R0451SKB003CTP, sodium solution100 mM50 mLBusiness supply
R0451SKB009CTP, sodium solution100 mM500 mLBusiness supply
R00016SKB002GTP, tris solution200 mM5 mLBusiness supply
R00016SKB005GTP, tris solution200 mM50 mLBusiness supply
R00016SKB004GTP, tris solution200 mM500 mLBusiness supply
R046SKB001GTP, sodium solution100 mM3.5 mLBusiness supply
R046SKB002GTP, sodium solution100 mM20.5 mLBusiness supply
R0461SKB003GTP, sodium solution100 mM50 mLBusiness supply
R0461SKB009GTP, sodium solution100 mM500 mLBusiness supply
R00017SKB005UTP, tris solution200 mM50 mLBusiness supply
R00017SKB004UTP, tris solution200 mM500 mLBusiness supply
R047SKB001UTP, sodium solution100 mM3.5 mLBusiness supply
R0471SKB006UTP, sodium solution100 mM20 mLBusiness supply
R0471SKB002UTP, sodium solution100 mM100 mLBusiness supply
R0471SKB003UTP, sodium solution100 mM500 mLBusiness supply
R00014SKB002CATP, Tris Solution200 mM5 mLInstant purchase
R044SKB001CATP, Sodium Solution100 mM3.5 mLInstant purchase
R00015SKB002CCTP, Tris Solution200 mM5 mLInstant purchase
R045SKB001CCTP, Sodium Solution100 mM3.5 mLInstant purchase
R00016SKB002CGTP, Tris Solution200 mM5 mLInstant purchase
R046SKB001CGTP, Sodium Solution100 mM3.5 mLInstant purchase
R00017SKB002CUTP, Tris Solution200 mM5 mLInstant purchase
R047SKB001CUTP, Sodium Solution100 mM3.5 mLInstant purchase
Catalog number R00017SKB002
Price (JPY)
-
Product Type:
UTP, tris solution
Concentration:
200 mM
Quantity:
5 mL
Format:
Business supply
Thermo Scientific TheraPure GMP nucleoside triphosphates (NTPs) are manufactured to ICH Q7 GMP principles and supported by a comprehensive documentation package. They are suitable as core building blocks for mRNA synthesis in the development and manufacturing of mRNA therapeutics. Nucleotides ATP, CTP, GTP, and UTP are available and are supplied in sodium- or Tris-titrated solutions.

Thermo Scientific TheraPure GMP* nucleoside triphosphates (NTPs) are manufactured to ICH Q7 GMP principles and supported by a comprehensive documentation package. They are suitable as core building blocks for mRNA synthesis in the development and manufacturing of mRNA therapeutics. Nucleotides ATP, CTP, GTP, and UTP are available and are supplied in sodium- or Tris-titrated solutions.

Designed for process development and manufacturing

TheraPure GMP nucleotides help accelerate mRNA vaccine and therapeutics production from preclinical development to commercialization by providing:

  • Quality—meet critical product quality needs to accelerate therapeutic development and manufacture
  • Minimal risk—reduce risks in developing a therapeutic by utilizing proven products
  • Technical partnership—partner with expert product and technical support
  • Secure and consistent supply—count on a secure and stable supply of product at a global scale

Supported by unique quality system

TheraPure GMP nucleotides are supported by a comprehensive quality system and documentation, including:

  • Manufactured to relevant ICH Q7 GMP guidelines
  • Animal-origin–free (AOF) manufacturing processes, materials, and facilities
  • Validated manufacturing processes and analytical test methods
  • Comprehensive impurity profiles
  • Verified compendial assays where applicable
  • Product stability data
  • Tested by analytical methods following ICH Q2 guidelines whenever possible
  • Drug master file (DMF)

Two available formats

These two formats are offered to accommodate different mRNA manufacturing processes:

  • 100 mM aqueous solution with Na+ titrated to pH 7.3–7.5 with NaOH
  • 200 mM aqueous solution with Tris counterion titrated to pH 7.3–7.7 with Tris base

Modified nucleotides

TheraPure GMP N1-methylpseudo-UTP is also available.

*TheraPure GMP refers to the quality level of the raw, ancillary, or starting materials to be used for further manufacturing. TheraPure GMP products are manufactured in facilities with ISO 9001–certified quality management systems that operate in accordance with relevant good manufacturing practice (GMP) principles, as outlined in ICH Q7 or equivalent guidance documents or standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorTransparent
For Use With (Application)IVT Transcription, RNA Synthesis, mRNA Synthesis
FormatBusiness supply
Label or DyeUnlabeled
Molecular Weight (g/mol)484.14 g/mol
Product TypeUTP, tris solution
Purity≥99%
Quantity5 mL
Shelf LifeUp to 36 months after manufacturing when stored at –20°C
Concentration200 mM
Product LineTheraPure™ GMP
pH7.3 to 7.7
Unit SizeEach