Aminoallyl-UTP Solution (50 mM)
Thermo Scientific™

Aminoallyl-UTP Solution (50 mM)

Thermo Scientific Aminoallyl-UTP (5-[3-aminoallyl]-2'-uridine-5'-triphosphate) is supplied as 50 mM aqueous solution titrated to pH 7.0 with NaOH.Aminoallyl-UTP can be enzymaticallyRead more
Have Questions?
Catalog NumberQuantity
R109150 μL
Catalog number R1091
Price (USD)
241.00
Each
Quantity:
50 μL
Request bulk or custom format
Price (USD)
241.00
Each

Thermo Scientific Aminoallyl-UTP (5-[3-aminoallyl]-2'-uridine-5'-triphosphate) is supplied as 50 mM aqueous solution titrated to pH 7.0 with NaOH.

Aminoallyl-UTP can be enzymatically incorporated into RNA with T7, T3 and SP6 RNA polymerases. The resulting amine-containing RNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin, or hapten.

Applications
• Indirect non-radioactive enzymatic labeling of RNA during in vitro transcription

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Non-Radioactive Labeling
Labeling MethodDirect Labeling
Label or DyeAminoallyl
Product TypeAminoallyl-UTP Solution
Quantity50 μL
Shipping ConditionDry Ice
Concentration50 mM
FormLiquid
Unit SizeEach
Contents & Storage
Aminoallyl-UTP Solution, 50 mM (50 μL)

Store at –20°C.