Product Image
Thermo Scientific™

Sensititre™ Myco RAPMYCOI AST Plate

Confidently perform susceptibility testing of rapidly growing non-tuberculosis mycobacteria with this convenient and cost-effective plate.

Have Questions?
Catalog NumberQuantity
RAPMYCOI10/Pk.
Catalog number RAPMYCOI
Price (MXN)
-
Quantity:
10/Pk.

Perform accurate susceptibility testing of rapidly growing non-tuberculous mycobacteria with Thermo Scientific™ Sensititre™ RAPMYCOI AST Plate. Test Mycobacterium spp. including rapid growing Mycobacterium fortuitum group or M. smegmatis group, Nocardia sp. and other aerobic actinomycetes with consistent endpoint determination (please note that a Nocardia RUO format, was also recently made available that follows updated guidelines issued by CLSI). Sensititre RAPMYCOI AST plate is intended for In Vitro Diagnostic Use.

Achieve reliable, consistent results for rapidly growing non-tuberculosis mycobacteria isolates with  Sensititre Myco Susceptibility Plate.

  • Extended shelf life, room-temperature storage eliminates inventory control concerns
  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Easy set-up procedures and results in three days for rapid-growing organisms
  • Test 15 antimicrobials on a single AST plate for cost-effective results.

The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.

  • Scalable – flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
  • Flexible – over 300 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
  • Tailor-made – design your own AST panel to meet specific formulary requirements and prescription protocols based on real-time trends and local epidemiology

Not all products are available for sale in all territories. Please inquire.

 

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
0.5 McFarland StandardH2O (Part No. T3339)
Antibiotic CodeVarious
Antimicrobial AgentsVarious
Broth TypeMHB w/ TES (Part No. T3462)
DescriptionSensititre Myco Susceptibility Plate- Rapid Growing
For Use With (Application)Rapid-growing Mycobacteria, Nocardia, Actinomycetes
Incubation ConditionsRapid-growing mycobacterium: 30°C O2. Nocardia and Actinomycetes: 35°C O2
Incubation TimeRapid-growing mycobacterium: 3-5 days. Nocardia and Actinomycetes: 2-3 days.
Isolates Per Plate1
No. per Pack10
Packaging TypeBox
Quantity10/Pk.
Reading OptionsManual
Transfer Volume50µL
Volume Per Well100 μL
Product TypeMIC Plate
SpeciesMycobacterium
Target Organism ClassRapidly-growing, Non-tuberculous Mycobacteria
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach